Learn More
Raybiotech Inc Human jun-D Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHJUND1
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical jun-D binding sequences to semi-quantitatively detect active jun-D in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0927