Learn More
Raybiotech Inc Human NF-κB p50 Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHP501
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical NF-κB binding sequences to semi-quantitatively detect active NF-κB in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0932