Learn More
Raybiotech Inc Human NRF2 Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHNRF21
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical NRF2 binding sequences to semi-quantitatively detect active NRF2 in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0930