Learn More
Raybiotech Inc Human PPAR-delta Transcription Factor Activity Assay.
Supplier:  Raybiotech Inc TFEHPPARD1
Encompass_Preferred
This assay uses a dsDNA coated plate with canonical PPAR-delta binding sequences to semi-quantitatively detect active PPAR-delta in lysates or nuclear extracts. Dry ice shipment (additional fee). Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments.
Catalog No. 50-195-0938