Learn More
REPLIGEN CORPORATION ELISA Kit for the Detection of NGL-Impact A Ligand
SDP
Supplier:  REPLIGEN CORPORATION 92221
Encompass
For downstream processes that utilize a Praesto Jetted A50 capture step, Repligen offers the only kit on the market that includes the NGL-Impact A Ligand standard for the most accurate quantitation.
Catalog No. NC1898463
 Restricted Item