Learn More
Biomatik Corporation Mouse Defensin Alpha 6, Paneth Cell Specific (DEFa6) ELISA Kit
Supplier:  Biomatik Corporation EKU08355
Encompass_Preferred
Mouse Defensin Alpha 6, Paneth Cell Specific (DEFa6) ELISA Kit for detecting DEFa6 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50704. Gene ID: 13240. Target Synonyms: DEF6; Defensin-6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3031