Learn More
Biomatik Corporation Human Alpha-2-Macroglobulin Like Protein 1 (a2ML1) ELISA Kit
Supplier:  Biomatik Corporation EKU02281
Encompass_Preferred
Human Alpha-2-Macroglobulin Like Protein 1 (a2ML1) ELISA Kit for detecting a2ML1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A8K2U0. Gene ID: 144568. Target Synonyms: CPAMD9; C3 And PZP-Like,Alpha-2-Macroglobulin Domain Containing 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2084