Learn More
Biomatik Corporation Human Angiogenic Factor With G Patch And FHA Domains 1 (AGGF1) ELISA Kit
Supplier:  Biomatik Corporation EKU02348
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8N302. Target Synonyms: GPATC7; GPATCH7; VG5Q; Angiogenic factor VG5Q; G patch domain-containing protein 7; Vasculogenesis gene on 5q protein. Restriction: For Research Use Only.
Catalog No. 50-229-2097