Learn More
Biomatik Corporation Mouse Angiogenin (ANG) ELISA Kit
Supplier:  Biomatik Corporation EKU02353
Encompass_Preferred
Mouse Angiogenin (ANG) ELISA Kit for detecting ANG in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.131ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21570. Gene ID: 11727. Target Synonyms: RNASE5; Ribonuclease,RNase A Family 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2099