Learn More
Biomatik Corporation Human Angiotensin II Receptor 2 (AGTR2) ELISA Kit
Supplier:  Biomatik Corporation EKU02411
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.056ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: AT2R; AT2-R; ATGR2; AT2; MRX88. Restriction: For Research Use Only.
Catalog No. 50-229-2108