Learn More
Biomatik Corporation Human Myeloperoxidase (MPO) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU02464
Encompass_Preferred
Human Myeloperoxidase (MPO) Anti-antigen ELISA Kit for detecting MPO in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P05164. Gene ID: 4353. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2116