Learn More
Biomatik Corporation Human Anti-Sperm Antibody (AsAb) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU02466
Encompass_Preferred
Human Anti-Sperm Antibody (AsAb) Anti-antigen ELISA Kit for detecting AsAb in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Target Synonyms: Anti-Spermatozoa Antibodies; Sperm Antibodies; Antisperm Antibodies. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2118