Learn More
Biomatik Corporation Rat Apolipoprotein E (APOE) ELISA Kit
Supplier:  Biomatik Corporation EKU02520
Encompass_Preferred
Rat Apolipoprotein E (APOE) ELISA Kit for detecting APOE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.92ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02650. Gene ID: 25728. Target Synonyms: Apo-E; AD2; Apoprotein; Alzheimer Disease 2 (E4-Associated,Late Onset. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2127