Learn More
Biomatik Corporation Rat Aquaporin 5 (AQP5) ELISA Kit
Supplier:  Biomatik Corporation EKU02550
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.125ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-2129