Learn More
Biomatik Corporation Mouse 5-Lipoxygenase (5-LO) ELISA Kit
Supplier:  Biomatik Corporation EKU02558
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P48999. Target Synonyms: LOX5; 5-LOX; ALOX-5; ALOX5; 5LPG; LOG5; Lipoxygenase 5. Restriction: For Research Use Only.
Catalog No. 50-229-2131