Learn More
Biomatik Corporation Rat Aryl Hydrocarbon Receptor (AhR) ELISA Kit
Supplier:  Biomatik Corporation EKU02577
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.102ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P41738. Target Synonyms: BHLHE76; Class E basic helix-loop-helix protein 76. Restriction: For Research Use Only.
Catalog No. 50-229-2133