Learn More
Biomatik Corporation Human Asialoglycoprotein Receptor 2 (ASGR2) ELISA Kit
Supplier:  Biomatik Corporation EKU02588
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 62.5-4,000pg/mL. Sensitivity: 23.3pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P07307. Target Synonyms: CLEC4H2; Asialoglyco Protein Receptor 2; C-type lectin domain family 4 member H2; Hepatic lectin H2. Restriction: For Research Use Only.
Catalog No. 50-229-2136