Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter C3 (ABCC3) ELISA Kit
Supplier:  Biomatik Corporation EKU02614
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O15438. Target Synonyms: ABC31; MLP2; MOATD; MRP3; cMOAT2; Canalicular Multispecific Organic Anion Transporter 2; Multi-specific organic anion transporter D; Multidrug resistance-associated protein 3. Restriction: For Research Use Only.
Catalog No. 50-229-2141