Learn More
Biomatik Corporation Rat BH3 Interacting Domain Death Agonist (Bid) ELISA Kit
Supplier:  Biomatik Corporation EKU02717
Encompass_Preferred
Rat BH3 Interacting Domain Death Agonist (Bid) ELISA Kit for detecting Bid in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JLT6. Gene ID: 64625. Target Synonyms: p22 BID. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2158