Learn More
Biomatik Corporation Bovine Bone Morphogenetic Protein 4 (BMP4) ELISA Kit
Supplier:  Biomatik Corporation EKU02751
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Bovine. Detection Range: 15.6-1,000pg/mL. Sensitivity: 6.3pg/m. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q2KJH1. Target Synonyms: BMP2B; BMP2-B; BMP2B1; ZYME; DVR4; Bone morphogenetic protein 2B. Restriction: For Research Use Only.
Catalog No. 50-229-2162