Learn More
Biomatik Corporation Carnosine (Car) ELISA Kit
Supplier:  Biomatik Corporation EKU02967
Encompass_Preferred
Carnosine (Car) ELISA Kit for detecting Car in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 11.11-900ng/ml. Sensitivity: 4.51ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: β-Alanyl-L-Histidine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2187