Learn More
Biomatik Corporation Mouse Caspase 8 (CASP8) ELISA Kit
Supplier:  Biomatik Corporation EKU02999
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.127ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O89110. Target Synonyms: CAP4; FLICE; MACH; MCH5; Cysteinyl Aspartate Specific Proteinases 8; Apoptotic cysteine protease; FADD-homologous ICE/ced-3-like protease; MORT1-associated ced-3 homolog. Restriction: For Research Use Only.
Catalog No. 50-229-2192