Learn More
Biomatik Corporation Mouse Chemokine C-C-Motif Ligand 3 Like Protein 1 (CCL3L1) ELISA Kit
Supplier:  Biomatik Corporation EKU03096
Encompass_Preferred
Mouse Chemokine C-C-Motif Ligand 3 Like Protein 1 (CCL3L1) ELISA Kit for detecting CCL3L1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 37pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q03366. Gene ID: 20306. Target Synonyms: CCL3-L1; LD78; SCYA3L; SCYA3L1; MIP1AP; MIP1-Alpha-P; Small Inducible Cytokine A3-Like 1; Macrophage Inflammatory Protein 1-Alpha-P; Tonsillar Lymphocyte LD78 Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2211