Learn More
Biomatik Corporation Mouse Chemokine C-C-Motif Receptor 7 (CCR7) ELISA Kit
Supplier:  Biomatik Corporation EKU03107
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P47774. Target Synonyms: CD197; CCR7; BLR2; CDw197; CMKBR7; EBI1; Epstein-Barr Virus Induced Protein 1; EBV-induced G-protein coupled receptor 1; MIP-3 beta receptor. Restriction: For Research Use Only.
Catalog No. 50-229-2215