Learn More
Biomatik Corporation Human Chemokine C-X-C-Motif Receptor 7 (CXCR7) ELISA Kit
Supplier:  Biomatik Corporation EKU03124
Encompass_Preferred
Human Chemokine C-X-C-Motif Receptor 7 (CXCR7) ELISA Kit for detecting CXCR7 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25106. Gene ID: 57007. Target Synonyms: ACKR3; RDC1; CMKOR1; GPR159; Chemokine Orphan Receptor 1; Atypical chemokine receptor 3; G-protein coupled receptor 159; G-protein coupled receptor RDC1 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2218