Learn More
Biomatik Corporation Mouse Cholinergic Receptor, Muscarinic 3 (CHRM3) ELISA Kit
Supplier:  Biomatik Corporation EKU03160
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: CHR-M3; HM3; M-AChRM3; Muscarinic Acetylcholine Receptor M3; Cholinergic Receptor,Muscarinic 3. Restriction: For Research Use Only.
Catalog No. 50-229-2220