Learn More
Biomatik Corporation Human Chymotrypsinogen B1 (CTRB1) ELISA Kit
Supplier:  Biomatik Corporation EKU03188
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.61ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P17538. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-2222