Learn More
Biomatik Corporation Rat Citrate Synthase (CS) ELISA Kit
Supplier:  Biomatik Corporation EKU03198
Encompass_Preferred
Rat Citrate Synthase (CS) ELISA Kit for detecting CS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VHF5. Gene ID: 170587. Target Synonyms: Citrate (Si)-synthase, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2226