Learn More
Biomatik Corporation Mouse Coagulation Factor II (F2) ELISA Kit
Supplier:  Biomatik Corporation EKU03257
Encompass_Preferred
Mouse Coagulation Factor II (F2) ELISA Kit for detecting F2 in serum. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8,000ng/ml. Sensitivity: 45ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19221. Gene ID: 14061. Target Synonyms: FII; TM; PT; Thrombin; Prothrombin; Pro-Thrombin; Activation peptide fragment 1; Activation peptide fragment 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2234