Learn More
Biomatik Corporation Rat Coagulation Factor X (F10) ELISA Kit
Supplier:  Biomatik Corporation EKU03274
Encompass_Preferred
Rat Coagulation Factor X (F10) ELISA Kit for detecting F10 in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63207. Gene ID: 29243. Target Synonyms: FX; FXA; Stuart Prower Factor; Thrombokinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2238