Learn More
Biomatik Corporation Human Coagulation Factor XIII B Polypeptide (F13B) ELISA Kit
Supplier:  Biomatik Corporation EKU03284
Encompass_Preferred
Human Coagulation Factor XIII B Polypeptide (F13B) ELISA Kit for detecting F13B in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 617.3-50,000ng/ml. Sensitivity: 243.7ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P05160. Gene ID: 2165. Target Synonyms: F13-B; FXIIIB; Fibrin-stabilizing factor B subunit; Protein-glutamine gamma-glutamyltransferase B chain; Transglutaminase B chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2240