Learn More
Biomatik Corporation Bovine Collagen Type III (COL3) ELISA Kit
Supplier:  Biomatik Corporation EKU03314
Encompass_Preferred
Bovine Collagen Type III (COL3) ELISA Kit for detecting COL3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.47-30ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04258. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2245