Learn More
Biomatik Corporation Human Collagen Type VIII Alpha 1 (COL8a1) ELISA Kit
Supplier:  Biomatik Corporation EKU03340
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P27658. Target Synonyms: COL8-A1; Collagen Alpha-1(VIII)chain; Endothelial collagen; Vastatin. Restriction: For Research Use Only.
Catalog No. 50-229-2248