Learn More
Biomatik Corporation Human Connexin 26 (CX26) ELISA Kit
Supplier:  Biomatik Corporation EKU03441
Encompass_Preferred
Human Connexin 26 (CX26) ELISA Kit for detecting CX26 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29033. Gene ID: 2706. Target Synonyms: KID; DFNA3; DFNB1; HID; NSRD1; PPK; GJB2; Gap Junction Protein Beta 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2257