Learn More
Biomatik Corporation Human Coordinator Of PRMT5 And Differentiation Stimulator (COPRS) ELISA Kit
Supplier:  Biomatik Corporation EKU03451
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: COPR5; TTP1; C17orf79; Cooperator Of PRMT5. Restriction: For Research Use Only.
Catalog No. 50-229-2259