Learn More
Biomatik Corporation Human CREB Binding Protein (CREBBP) ELISA Kit
Supplier:  Biomatik Corporation EKU03497
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.112ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q92793. Target Synonyms: CBP; RSTS; RTS; KAT3A; Rubinstein-Taybi Syndrome. Restriction: For Research Use Only.
Catalog No. 50-229-2263