Learn More
Biomatik Corporation Human Cyclin Dependent Kinase 5 (CDK5) ELISA Kit
Supplier:  Biomatik Corporation EKU03539
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.106ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q00535. Target Synonyms: PSSALRE; CDKN5; Cell division protein kinase 5; Serine/threonine-protein kinase PSSALRE; Tau protein kinase II catalytic subunit;TPKII catalytic subunit. Restriction: For Research Use Only.
Catalog No. 50-229-2272