Learn More
Biomatik Corporation Human Cysteine Conjugate Beta Lyase, Cytoplasmic (CCbL1) ELISA Kit
Supplier:  Biomatik Corporation EKU03571
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 78.12-5,000pg/mL. Sensitivity: 27pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q16773. Target Synonyms: KATI; GTK; Cysteine-S-conjugate beta-lyase; Glutamine-phenylpyruvate transaminase; Kyneurenine Aminotransferase; Kynurenine Oxoglutarate Transaminase. Restriction: For Research Use Only.
Catalog No. 50-229-2280