Learn More
Biomatik Corporation Human Dopamine Beta Hydroxylase (DbH) ELISA Kit
Supplier:  Biomatik Corporation EKU03780
Encompass_Preferred
Human Dopamine Beta Hydroxylase (DbH) ELISA Kit for detecting DbH in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09172. Gene ID: 1621. Target Synonyms: DBM; Dopamine Beta-Monooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2319