Learn More
Biomatik Corporation Human E1A Binding Protein P300 (EP300) ELISA Kit
Supplier:  Biomatik Corporation EKU03818
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q09472. Target Synonyms: KAT3B; p300; Histone Acetyltransferase p300. Restriction: For Research Use Only.
Catalog No. 50-229-2328