Learn More
Biomatik Corporation Human E2F Transcription Factor 1 (E2F1) ELISA Kit
Supplier:  Biomatik Corporation EKU03821
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: RBBP3; RBP3; RBAP-1; Retinoblastoma-associated protein 1; Retinoblastoma-binding protein 3; pRB-binding protein E2F-1. Restriction: For Research Use Only.
Catalog No. 50-229-2330