Learn More
Biomatik Corporation Human Elastase 3B (ELA3B) ELISA Kit
Supplier:  Biomatik Corporation EKU03848
Encompass_Preferred
Human Elastase 3B (ELA3B) ELISA Kit for detecting ELA3B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08861. Gene ID: 23436. Target Synonyms: CELA3B; Chymotrypsin Like Elastase Family Member 3B; Protease E; Fecal Elastase; Elastase 1; Cholesterol-Binding Pancreatic Protease; Pancreatic Endopeptidase E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2335