Learn More
Biomatik Corporation Human Endoplasmic Reticulum Aminopeptidase 2 (ERAP2) ELISA Kit
Supplier:  Biomatik Corporation EKU03876
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q6P179. Target Synonyms: L-RAP; LRAP; Leukocyte-Derived Arginine Aminopeptidase. Restriction: For Research Use Only.
Catalog No. 50-229-2339