Learn More
Biomatik Corporation Mouse Enhancer Of Zeste Homolog 1 (EZH1) ELISA Kit
Supplier:  Biomatik Corporation EKU03898
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.061ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: KMT6B; ENX-2; Histone-lysine N-methyltransferase EZH1. Restriction: For Research Use Only.
Catalog No. 50-229-2342