Learn More
Biomatik Corporation Human Epithelial Cell Adhesion Molecule (EPCAM) ELISA Kit
Supplier:  Biomatik Corporation EKU03949
Encompass_Preferred
Human Epithelial Cell Adhesion Molecule (EPCAM) ELISA Kit for detecting EPCAM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16422. Gene ID: 4072. Target Synonyms: CD326; TACSTD1; EGP; KSA; M4S1; MIC18; TROP1; HEGP2; ESA; Epithelial glycoprotein; Adenocarcinoma-associated antigen; Tumor Associated Calcium Signal Transducer 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2353