Learn More
Biomatik Corporation Estriol (E3) ELISA Kit
Supplier:  Biomatik Corporation EKU03972
Encompass_Preferred
Estriol (E3) ELISA Kit for detecting E3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 1.23-100ng/ml. Sensitivity: 0.47ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: oestriol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2357