Learn More
Biomatik Corporation Human Fatty Acid Desaturase 2 (FADS2) ELISA Kit
Supplier:  Biomatik Corporation EKU04049
Encompass_Preferred
Human Fatty Acid Desaturase 2 (FADS2) ELISA Kit for detecting FADS2 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95864. Gene ID: 9415. Target Synonyms: LLCDL2; FADSD6; D6D; TU13; DES6; Delta-6-Desaturase; Delta (6) fatty acid desaturase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2369