Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 13 (FGF13) ELISA Kit
Supplier:  Biomatik Corporation EKU04137
Encompass_Preferred
Mouse Fibroblast Growth Factor 13 (FGF13) ELISA Kit for detecting FGF13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70377. Gene ID: 14168. Target Synonyms: FHF2; Fibroblast Growth Factor Homologous Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2379