Learn More
Biomatik Corporation Mouse Fibulin 7 (FBLN7) ELISA Kit
Supplier:  Biomatik Corporation EKU04215
Encompass_Preferred
Mouse Fibulin 7 (FBLN7) ELISA Kit for detecting FBLN7 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q501P1. Gene ID: 70370. Target Synonyms: TM14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2389