Learn More
Biomatik Corporation Rat Forkhead Box P3 (FOXP3) ELISA Kit
Supplier:  Biomatik Corporation EKU04264
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: FOX-P3; AIID; DIETER; IPEX; JM2; PIDX; XPID; SCURFIN; Forkhead Box Protein P3; Immune Dysregulation,Polyendocrinopathy,Enteropathy,X-Linked. Restriction: For Research Use Only.
Catalog No. 50-229-2398